Alicat MC-Series Mass Flow Controllers, SLPM, Large Pneutronics Valve
Provides accurate, real-time mass and volumetric flow readings
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • Lifetime warranty
These large pneutronics valve mass flow controllers, featuring a fast control response time of just 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They are capable of measuring and controlling flow generated by both positive pressure and suction. Real-time monitoring of flow, pressure, and temperature is available directly on the display screen.
The controllers come with 98 pre-loaded gas calibrations, which can be seamlessly switched while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure, with flow rates displayed in standard liters per minute (SLPM). Additional key features include an LCD display with backlight, a data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flow Controller with Display, Large Pneutronics Valve, 0 - 50 SLPM 1530909 Mfr # MCP-50SLPM-D
COMPARE
50 ± 0.4% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$3,138.32CAD / Each
Alicat Mass Flow Controller with Display, Large Pneutronics Valve, 0 - 100 SLPM 1530910 Mfr # MCP-100SLPM-D
COMPARE
100 ± 0.4% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$3,138.32CAD / Each
Alicat Mass Flow Controller with Display, Large Pneutronics Valve, 0 - 250 SLPM 1530911 Mfr # MCP-250SLPM-D
COMPARE
250 ± 0.4% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$3,963.42CAD / Each
Alicat Mass Flow Controller with Display, LRV-67x51, 0 - 500 SLPM 1530912 Mfr # MCR-500SLPM-D
COMPARE
500 ± 0.4% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$3,735.04CAD / Each
Alicat Mass Flow Controller with Display, Low Drop Valve, 0 - 1000 SLPM 1530913 Mfr # MCR-1000SLPM-D
COMPARE
1000 ± 0.8% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$4,957.96CAD / Each
Alicat Mass Flow Controller with  Display, Parallel Wound Coil, 0 - 2000 SLPM 1530914 Mfr # MCR-2000SLPM-D-PAR
COMPARE
2000 ± 0.8% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$6,630.26CAD / Each
Alicat Mass Flow Controller with Display, LRV, Parallel Wound Coil, 0 - 3000 SLPM 1530915 Mfr # MCR-3000SLPM-D-PAR
COMPARE
3000 ± 0.8% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$7,219.61CAD / Each
Alicat Mass Flow Controller with  Display, Dual Parallel Rolamite Valves, 0 - 5000 SLPM 1530916 Mfr # MCRH-5000SLPM-D-PAR
COMPARE
5000 ± 0.8% of reading and ± 0.2% of full scale ± 0.2% of reading and ± 0.02% of full scale
$10,917.82CAD / Each
Showing 1- 8 of 8