Alicat M-Series Mass Flowmeters with Digital Display, SCCM
Provides accurate, real-time mass and volumetric flow readings
  • No warm-up required
  • Ninety-eight pre-loaded gas calibrations
  • Ten millisecond measurement response time
  • NIST-traceable calibration certificate included - accuracy up to ±0.5% of reading
  • Lifetime warranty
These mass flowmeters, with a fast control response time of 30 milliseconds (ms), offer quick integration into test stands for accurate verification without any warm-up time. They come with 98 pre-loaded gas calibrations, which can be switched seamlessly while maintaining accuracy. The stand-alone units measure mass flow, volumetric flow, temperature, and absolute pressure with flow rates displayed in standard cubic centimeters per minute (SCCM). Key features include a backlit monochrome display, data I/O port for logging data to your computer, and flexible mounting options, including upside-down installation.
Custom Electrical connections include: 6-pin locking, 8-pin M12, 9-pin DB-9, 15-pin DB-15. Please contact [email protected] if you require an electrical connection other than the 8-pin mini-DIN default.
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (SCCM) Accuracy Repeatability
Availability Pricing
Alicat Mass Flowmeter with Display, 0 - 1 SCCM 1530872 Mfr # M-1SCCM-D
COMPARE
1 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$2,806.81CAD / Each
Alicat Mass Flowmeter with Display, 0 - 2 SCCM 1530873 Mfr # M-2SCCM-D
COMPARE
2 ±0.8% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$2,887.84CAD / Each
Alicat Mass Flowmeter with Display, 0 - 5 SCCM 1530874 Mfr # M-5SCCM-D
COMPARE
5 ±0.4% of reading + 0.2% of full scale ±0.2% of reading + 0.02% of full scale
$1,966.98CAD / Each
Alicat Mass Flowmeter with Display, 0 - 10 SCCM 1530875 Mfr # M-10SCCM-D
COMPARE
10 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$1,966.98CAD / Each
Alicat Mass Flowmeter with Display, 0 - 20 SCCM 1530876 Mfr # M-20SCCM-D
COMPARE
20 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$1,966.98CAD / Each
Alicat Mass Flowmeter with Display, 0 - 50 SCCM 1530877 Mfr # M-50SCCM-D
COMPARE
50 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$1,966.98CAD / Each
Alicat Mass Flowmeter with Display, 0 - 100 SCCM 1530878 Mfr # M-100SCCM-D
COMPARE
100 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$1,966.98CAD / Each
Alicat Mass Flowmeter with Display, 0 - 200 SCCM 1530879 Mfr # M-200SCCM-D
COMPARE
200 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$2,033.28CAD / Each
Alicat Mass Flowmeter with Display, 0 - 500 SCCM 1530880 Mfr # M-500SCCM-D
COMPARE
500 ±0.6% of reading ±0.1% of reading + 0.02% of full scale
$1,966.98CAD / Each
Showing 1- 9 of 9