Advantec Sanitary Stainless Steel In-Line Filter Holders
Non-threaded sanitary inlet and outlet connections easily disassembled for thorough cleaning
  • Clamp hinges on one point
These sanitary stainless steel holders are for liquid and gas applications. Use for point-of-use filtration on sterile filling machines and for lab or pilot scale sterile filtration applications.
Body and support screen are 304 SS with PTFE gasket and FPM/silicone O-rings. Max pressure at inlet is 71 psi (4.9 bar).
$833.94 - $1,606.00CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm) Inlet Connection
Availability Pricing
Advantec 357000 Sanitary Stainless Steel In-Line Filter Holder, for 25 mm membranes 0664351 Mfr # 17307000
COMPARE
Stainless Steel 25 Hose barb (6mm)
$833.94CAD / Each
Advantec 357200 Sanitary Stainless Steel In-Line Filter Holder, for 47 mm membranes 0664353 Mfr # 17307200
COMPARE
Stainless Steel 47 Hose barb (8mm)
$1,606.00CAD / Each