Advantec Grade No. 5A Ashless Filter Paper for Coarse / Gelatinous Retention
Manufactured to exacting standards for uniform performance
  • Fast flow rate, retains coarse particulates and gelatinous precipitates (>10 m); filter hydroxides and metallic aerosols, environmental monitoring, silica in steel
Ideal for gravimetric analysis and environmental monitoring. These papers are made of the highest quality alpha cotton cellulose with low ash content. Papers are double acid washed in hydrochloric acid followed by hydroflouric acid, then rinsed with ultrapure water to neutralize. Sold in packs of 100.
$37.28 - $179.02CAD / Pkg of 100
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (cm) Material
Availability Pricing
Advantec N05A5.5CM Grade 5A Ashless Quantitative Filters, >10 µm, 5.5 cm; 100/Pk 8105080 Mfr # N05A5.5CM
COMPARE
5.5 Alpha Cotton Cellulose
$37.28CAD / Pkg of 100
Advantec N05A7.0CM Grade 5A Ashless Quantitative Filters, >10 µm, 7 cm; 100/Pk 8105082 Mfr # N05A7.0CM
COMPARE
7 Alpha Cotton Cellulose
$43.47CAD / Pkg of 100
Advantec N05A11.0CM Grade 5A Ashless Quantitative Filters, >10 µm, 11 cm; 100/Pk 8105088 Mfr # N05A11.0CM
COMPARE
11 Alpha Cotton Cellulose
$85.46CAD / Pkg of 100
Advantec N05A12.5CM Grade 5A Ashless Quantitative Filters, >10 µm, 12.5 cm; 100/Pk 8105090 Mfr # N05A12.5CM
COMPARE
12.5 Alpha Cotton Cellulose
$94.15CAD / Pkg of 100
Advantec N05A15.0CM Grade 5A Ashless Quantitative Filters, >10 µm, 15 cm; 100/Pk 8105092 Mfr # N05A15.0CM
COMPARE
15 Alpha Cotton Cellulose
$113.89CAD / Pkg of 100
Advantec N05A18.5CM Grade 5A Ashless Quantitative Filters, >10 µm, 18.5 cm; 100/Pk 8105110 Mfr # N05A18.5CM
COMPARE
18.5 Alpha Cotton Cellulose
$179.02CAD / Pkg of 100