Advantec Extraction Thimbles, Cellulose
Seamless, high-purity filters for extraction and air monitoring
Thimble filters are used in soxhlet apparatus for dust sampling and for extracting soluble components using organic solvents. Thimbles are made of cellulose. Typical applications include analysis of fats, oils, grease, organics, pesticides, pollutants, and additives in plastic/rubber materials.
Cellulose thimbles are suitable for extraction of organic compounds and dust sampling. Lipid content is <0.1% by weight; thickness is 1.5 mm.
$173.86 - $304.62CAD / Pkg of 25
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Height (mm) Material
Availability Pricing
Advantec NO8422X80MM Extraction Thimbles, Cellulose, 22 mm ID x 80 mm H; 25/pack 2982100 Mfr # N08422X25X80MM
COMPARE
22 80 Cellulose
$186.38CAD / Pkg of 25
Advantec N08425X60MM Extraction Thimbles, Cellulose, 25 mm ID x 60 mm H; 25/pack 2982102 Mfr # N08425X29X60MM
COMPARE
25 60 Cellulose
$194.49CAD / Pkg of 25
Advantec N08433X80MM Extraction Thimbles, Cellulose, 33 mm ID x 80 mm H; 25/pack 2982104 Mfr # N08433X37X80MM
COMPARE
33 80 Cellulose
$197.80CAD / Pkg of 25
Advantec NO8443X123MM Extraction Thimbles, Cellulose, 43 mm ID x 123 mm H; 25/pack 2982106 Mfr # N08443X47X123MM
COMPARE
43 123 Cellulose
$304.62CAD / Pkg of 25
Advantec N08422X65MM Extraction Thimbles, Cellulose, 22 mm ID x 65 mm H; 25/pack 2982108 Mfr # N08422X25X65MM
COMPARE
22 65 Cellulose
$173.86CAD / Pkg of 25
Advantec N08425X90MM Extraction Thimbles, Cellulose, 25 mm ID x 90 mm H; 25/pack 2982110 Mfr # N08425X29X90MM
COMPARE
25 90 Cellulose
$186.02CAD / Pkg of 25
Advantec N08430X100MM Extraction Thimbles, Cellulose, 30 mm ID x 100 mm H; 25/pack 2982112 Mfr # N08430X34X100MM
COMPARE
30 100 Cellulose
$189.33CAD / Pkg of 25
Advantec N08430X80MM Extraction Thimbles, Cellulose, 30 mm ID x 80 mm H; 25/pack 2982114 Mfr # N08430X34X80MM
COMPARE
30 80 Cellulose
$198.17CAD / Pkg of 25
Advantec N08419X90MM Extraction Thimbles, Cellulose, 19 mm ID x 90 mm H; 25/pack 2982144 Mfr # N08419X22X90MM
COMPARE
19 90 Cellulose
$173.86CAD / Pkg of 25
Advantec N08425X80MM Extraction Thimbles, Cellulose, 25 mm ID x 80 mm H; 25/pack 2982146 Mfr # N08425X29X80MM
COMPARE
25 80 Cellulose
$194.49CAD / Pkg of 25
Advantec N08425X100MM Extraction Thimbles, Cellulose, 25 mm ID x 100 mm H; 25/pack 2982148 Mfr # N08425X29X100MM
COMPARE
25 100 Cellulose
$186.02CAD / Pkg of 25
Advantec N08428X100MM Extraction Thimbles, Cellulose, 28 mm ID x 100 mm H; 25/pack 2982150 Mfr # N08428X32X100MM
COMPARE
28 100 Cellulose
$188.96CAD / Pkg of 25
Advantec N08433X100MM Extraction Thimbles, Cellulose, 33 mm ID x 100 mm H; 25/pack 2982152 Mfr # N08433X37X100MM
COMPARE
33 100 Cellulose
$206.64CAD / Pkg of 25
Showing 1- 13 of 13