Advantec Disposable Capsule Filters
Polypropylene (PP) housing available with PTFE or PES membrane, or PP depth media
  • All-polypropylene housing is thermally fused and offers broad chemical compatibility; diameter is a standard 78 mm
  • Capsules are sterilizable by autoclave or ethylene oxide (EtO);
  • Max operating pressure is 57 psi (3.9 bar); max operating temperature is 140°F (60°C)
  • Capsules are vented at both ends for bleeding trapped air and for draining
$133.78 - $561.36CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Effective Filtration Area (cm²) Membrane Material Pore Size (micron)
Availability Pricing
Advantec CCS-020-C1B Disposable Capsule Filters, PES; 0.20 µm; 450 cm2; EA 2970201 Mfr # CCS-020-C1B
COMPARE
450 Polyethersulfone 0.2
$187.12CAD / Each
Advantec CCS-045-C1B Disposable Capsule Filters, PES; 0.45 µm; 450 cm2; EA 2970202 Mfr # CCS-045-C1B
COMPARE
450 Polyethersulfone 0.45
$196.33CAD / Each
Advantec CCS-020-D1B Disposable Capsule Filters, PES; 0.20 µm; 900 cm2; EA 2970203 Mfr # CCS-020-D1B
COMPARE
900 Polyethersulfone 0.2
$279.94CAD / Each
Advantec CCS-045-D1B Disposable Capsule Filters, PES; 0.45 µm; 900 cm2; EA 2970204 Mfr # CCS-045-D1B
COMPARE
900 Polyethersulfone 0.45
$267.42CAD / Each
Advantec CCS-020-D1HS Disposable Capsule Filters, PES; 0.20 µm; 900 cm2; presterilized; EA 2970205 Mfr # CCS-020-D1HS
COMPARE
900 Polyethersulfone 0.2
$335.93CAD / Each
Advantec CCP-JX-C1H Disposable Capsule Filters, Polypropylene Depth-Media; 1.0 µm; 500 cm2 2970211 Mfr # CCP-JX-C1H
COMPARE
500 Polypropylene 1
$154.34CAD / Each
Advantec CCP-1-C1H Disposable Capsule Filters, Polypropylene Depth-Media; 2-5 µm; 500 cm2 2970212 Mfr # CCP-1-C1H
COMPARE
500 Polypropylene 2-5
$133.78CAD / Each
Advantec CCP-JX-D1H Disposable Capsule Filters, Polypropylene Depth-Media; 1.0 µm; 1000 cm2 2970213 Mfr # CCP-JX-D1H
COMPARE
1000 Polypropylene 1
$196.33CAD / Each
Advantec CCP-1-D1H Disposable Capsule Filters, Polypropylene Depth-Media; 2-5 µm; 1000 cm2 2970214 Mfr # CCP-1-D1H
COMPARE
1000 Polypropylene 2-5
$196.33CAD / Each
Advantec CCP-3-D1H Disposable Capsule Filters, Polypropylene Depth-Media; 5-10 µm; 1200 cm2 2970216 Mfr # CCP-3-D1H
COMPARE
1200 Polypropylene 5-10
$187.12CAD / Each
Advantec CCF-020-C1H Disposable Capsule Filters, PTFE Hydrophobic; 0.20 µm; 570 cm2 2970221 Mfr # CCF-020-C1H
COMPARE
570 PTFE Hydrophobic 0.2
$267.42CAD / Each
Advantec CCF-100-C1H Disposable Capsule Filters, PTFE Hydrophobic; 1.0 µm; 570 cm2 2970222 Mfr # CCF-100-C1H
COMPARE
570 PTFE Hydrophobic 1
$267.42CAD / Each
Advantec CCF-A10-D1H Disposable Capsule Filters, PTFE Hydrophobic; 0.1 µm; 1150 cm2 2970223 Mfr # CCF-A10-D1H
COMPARE
1150 PTFE Hydrophobic 0.1
$561.36CAD / Each
Advantec CCF-020-D1H Disposable Capsule Filters, PTFE Hydrophobic; 0.20 µm; 1150 cm2 2970224 Mfr # CCF-020-D1H
COMPARE
1150 PTFE Hydrophobic 0.2
$391.92CAD / Each
Advantec CCF-100-D1H Disposable Capsule Filters, PTFE Hydrophobic; 1.0 µm; 1150 cm2 2970226 Mfr # CCF-100-D1H
COMPARE
1150 PTFE Hydrophobic 1
$374.24CAD / Each
Showing 1- 15 of 15