Adam Equipment Highland® Portable Balances
Compact, durable balances offer fast, accurate weighing
  • Weighing modes include parts counting, percent weighing, and accumulation of weight
  • RS-232 and USB interfaces provide connection to a computer or printer
  • Solid ABS housing and 304 stainless steel pan offer durability
  • Both internal and external calibration available to ensure balance accuracy
  • Large, backlit LCD is easy to read
  • Capacity tracker indicates used and remaining capacity
  • Dual tare keys accommodate both left- and right-handed users
  • Below-balance weighing hook enables density and specific gravity measurements
  • Internal rechargeable battery for convenient portable operation
Durable yet lightweight Highland® portable balances are packed with practical features for use in a lab or in the field, and in various educational and industrial applications. Easily calibrate the balances using the HandiCal® internal calibration feature; additional external calibration can be done using multiple weights.
Easily view measurements on an 18-mm high, 6-digit backlit LCD. Programmable backlight can be set to “always stay on”, “always off”, or “light only when weighing”. Low-battery indicator lights when battery needs charging; auto power-off feature conserves battery life. An audible alarm sounds when capacity is exceeded to help protect the balance from overload damage. Secure balance in place using a built-in security slot for a Kensington-type lock and cable (order separately).
$906.13 - $1,326.05CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Description
Availability Pricing
Adam Equipment Highland® Portable Precision Balance with Draft Shield, 120g x 0.001g 1171214 Mfr # HCB 123
COMPARE
120 0.001 Highland® Portable Precision Balance with Draft Shield, 120g x 0.001g
$1,046.11CAD / Each
Adam Equipment HCB153 Highland® Portable Precision Balance with Draft Shield, 150g x 0.005g 1171215 Mfr # HCB 153
COMPARE
150 0.005 Highland® Portable Precision Balance with Draft Shield, 150g x 0.005g
$906.13CAD / Each
Adam Equipment HCB302 Highland® Portable Precision Balance with Draft Shield, 300g x 0.01g 1171216 Mfr # HCB 302
COMPARE
300 0.01 Highland Portable Balance with Draft Shield, 300g x 0.01g
$906.13CAD / Each
Adam Equipment HCB 602H Highland® Portable Precision Balance with Draft Shield, 600g x 0.01g 1171217 Mfr # HCB 602H
COMPARE
600 0.01 Highland® Portable Precision Balance with Draft Shield, 600g x 0.01g
$1,134.51CAD / Each
Adam Equipment HCB602 Highland® Portable Precision Balance with Draft Shield, 600g x 0.02g 1171218 Mfr # HCB 602
COMPARE
600 0.02 Highland® Portable Precision Balance with Draft Shield, 600g x 0.02g
$906.13CAD / Each
Adam Equipment Highland® Portable Precision Balance with Draft Shield, 1000g x 0.01g 1171219 Mfr # HCB 1002
COMPARE
1000 0.01 Highland Portable Balance with Draft Shield, 1000g x 0.01g
$1,326.05CAD / Each
Adam Equipment Highland® Portable Precision Balance with Draft Shield, 1500g x 0.05g 1171220 Mfr # HCB 1502
COMPARE
1500 0.05 Highland® Portable Precision Balance with Draft Shield, 1500g x 0.05g
$1,053.47CAD / Each
Adam Equipment Highland® Portable Precision Balance with Draft Shield, 3000g x 0.1g 1171221 Mfr # HCB 3001
COMPARE
3000 0.1 Highland® Portable Precision Balance with Draft Shield, 3000g x 0.1g
$906.13CAD / Each
Adam Equipment HCB 6001 Highland® Portable Precision Balance, 6000g x 0.1g 5500118 Mfr # HCB 6001
COMPARE
6000 0.1 Highland® Portable Precision Balance, 6000g x 0.1g
$1,259.75CAD / Each
Showing 1- 9 of 9