Adam Equipment GC Industrial Scale Indicator
Compact counting indicator
  • Weighing
  • Counting
  • Preset counting
  • Bi-directional RS-232 to print out weight, average piece weight and number of pieces counted
  • Freely selectable sample size through keypad entry
  • Entry for average piece weight of an item to be counted
  • Can be linked with 1 to 4 loadcells
  • Wall-mount bracket
As a fully featured counting indicator, the GC offers a numeric keypad for unit weight and sample entry, and a large backlit display to show unit weight, total weight and number of pieces. With valuable features like check counting included, the GC offers the ability to warn the user when
a pre-defined count has been reached.
A solid ABS plastic housing and wall-mounted bracket makes the GC a durable and versatile indicator. The GC can be used in remote locations without power due to the internal rechargeable battery. It can also accumulate count totals through its memory facility.
$884.03CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Power (VAC) Description
Availability Pricing
Adam Equipment GC INDICATOR Counting Scale Indicator with Numeric Keypad and Three Displays 1171054 Mfr # GCa
COMPARE
1 x Lead Acid (Included) 115 Counting Scale Indicator with Numeric Keypad and Three Displays
$884.03CAD / Each
Adam Equipment GC INDICATOR (220V) Counting Scale Indicator with Numeric Keypad and Three Displays 220V 1171055 Mfr # GC
COMPARE
1 x Lead Acid (Included) 230 Counting Scale Indicator with Numeric Keypad and Three Displays 220V
$884.03CAD / Each