Adam Equipment Equinox Semi-Micro Balances
Remarkable precision for detailed testing
  • Intuitive, color, 5-inch (125 mm) touchscreen display provides high usability and visibility
  • Perform statistics on up to 500 consecutive weighs
  • Store and recall 3000 data points internally, or add a USB flash drive to expand capacity further
  • Time and date stamp meets Good Documentation Practices (GDP) requirements
  • High functionality includes checkweighing, parts counting, percentage weighing, and statistical analysis
The Equinox line of semi-micro balances provide many features and functions—at an affordable price. Take advantage of the dual-range capacity—higher resolution for your smaller samples!
Easily view results on the 5" (125 mm) display. This color touchscreen also makes function selection simple, and menu navigation is intuitive. Order the optional in-use cover to protect the screen, while still allowing operation.
The spirit-filled level indicator and adjustable feet ensures proper set up to obtain precise measurements. Perform checkweighing, parts counting, percent weighing, density determinations, dynamic weighing, formulation dosing, and statistical analysis.
$9,473.90 - $11,050.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity Dual Range Description
Availability Pricing
Adam Equipment EAB 125i Equinox Semi-Micro Balance, 62g x .01mg/120g x.1mg, Internal Calibration 1112501 Mfr # EAB 125i
COMPARE
62g / 120g Equinox Semi-Micro Balance, 62g x .01mg/120g x.1mg, Internal Calibration
$9,473.90CAD / Each
Adam Equipment EAB 225i Equinox Semi-Micro Balance, 82g x .01mg/220g x 0.1mg, Internal Calibration 1112502 Mfr # EAB 225i
COMPARE
82g / 220g Equinox Semi-Micro Balance, 82g x .01mg/220g x 0.1mg, Internal Calibration
$11,050.43CAD / Each