Adam Equipment AELP Industrial Pallet Beam Scales
Get fast, accurate pallet weighing results
  • Low-profile beams allow easy weighing of pallets
  • Lightweight with built-in wheels and handles for portability
  • Precision load cells for accurate weighing results
The AELP industrial pallet beam scales are ideal for weighing pallets using pallet trucks. Heavy-duty steel beams with OIML-approved load cells can be connected to either the AE 402 or GK weighing indicator.
$2,143.78 - $2,688.94CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
Adam Equipment AELP 1000 Pallet Beam Scale, 1000 kg x 0.2 kg 1111901 Mfr # AELP 1000
COMPARE
1000 2200 Pallet Beam Scale, 1000 kg x 0.2 kg
$2,143.78CAD / Each
Adam Equipment AELP 1000 Pallet Beam Scale with AE403 Indicator, 1000 kg x 0.2 kg, Universal Voltage 1111902 Mfr # AELP 1000 [AE403a]
COMPARE
1000 2200 Pallet Beam Scale with AE403 Indicator, 1000 kg x 0.2 kg, Universal Voltage
$2,688.94CAD / Each
Adam Equipment AELP 2000 Pallet Beam Scale, 2000 kg x 0.5 kg 1111904 Mfr # AELP 2000
COMPARE
2000 4400 Pallet Beam Scale, 2000 kg x 0.5 kg
$2,143.78CAD / Each
Adam Equipment AELP 2000 Pallet Beam Scale with AE403 Indicator, 2000 kg x 0. 5kg, Universal Voltage 1111906 Mfr # AELP 2000 [AE403a]
COMPARE
2000 4400 Pallet Beam Scale with AE403 Indicator, 2000 kg x 0. 5kg, Universal Voltage
$2,143.78CAD / Each
Adam Equipment AELP 1000 [AE403]-220V Pallet Beam Scale with AE403 Indicator, 1000 kg x 0.2 kg, 220 V 1111908 Mfr # AELP 1000 [AE403]-220V
COMPARE
1000 2200 Pallet Beam Scale with AE402 Indicator, 1000 kg x 0.2 kg, 220 V
$2,688.94CAD / Each
Adam Equipment AELP 2000 [AE403-220V Pallet Beam Scale with AE403 Indicator, 2000 kg x 0.5 kg, 220 V 1111911 Mfr # AELP 2000 [AE403-220V
COMPARE
2000 4400 Pallet Beam Scale with AE402 Indicator, 2000 kg x 0.5 kg, 220 V
$2,688.94CAD / Each