A&D Weighing HW-WP and HV-WP Series Washdown Industrial Scales
Water-resistant stainless steel construction offers protection from most challenging washdown environments
  • Bright traffic-style 5-step LED comparator lights provide accurate check-weighing
  • Ten-digit alphanumeric keypad with key lock and hold functions simplifies and speeds limit setting
  • Adjustable and detachable display with stainless steel bezel
  • IP-65 rated withstands harsh washdown and outdoor conditions
  • One-inch character LCD display with white backlight offers easy viewing even in dimly lit work areas
  • Counting function for inventory control
  • Works with A&D WinCT (data capture and graphing utility) for easy transfer to Excel
These scales provide counting, percentage, and comparator functions. Ideal for packaging, shipping, pharmaceutical, and industrial applications. Options include bidirectional USB and RS-232C interface for connection to printer or PC and comparator buzzer.
HW-CWP series of scales has high resolution of 1/10,000 or 1/12,000 depending on model.
$2,276.39 - $3,248.82CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing HW-10KCWP High Resolution Washdown Industrial Scale; Capacity 20 lb x 0.002lb 1112214 Mfr # HW-10KCWP
COMPARE
10 20 115
$2,843.64CAD / Each
A&D Weighing HW-60K-WP High Resolution Washdown Industrial Scale, Capacity 150 lb x 0.01 lb 1112220 Mfr # HW-60KCWP
COMPARE
60 150 115
$2,946.78CAD / Each
A&D Weighing HW-100KCWP High Resolution Washdown Industrial Scale, Capacity 200 lb 1112222 Mfr # HW-100KCWP
COMPARE
100 200 115
$3,094.12CAD / Each
A&D Weighing HW-200K-WP High Resolution Washdown Industrial Scale; Capacity 500 lb x 0.05 lb 1112226 Mfr # HW-200KCWP
COMPARE
220 500 115
$3,248.82CAD / Each
A&D Weighing HV-15KWP Tripple Resolution Washdown Industrial Scale; 6 lb/15 lb/30 lb 1122991 Mfr # HV-15KCWP
COMPARE
13.6 30 115
$2,276.39CAD / Each