A&D Weighing HV-C/CP and HW-C/CP Series Bench Scales
Multicolored comparator lights ensure accurate checkweighing
  • Accumulation function displays sum total of separate weighing results
  • Ten-digit keypad simplifies setting comparator limits
  • IP65-rated base and stainless steel pan allow for wet weighing and easy cleanup
  • Built-in key lock prevents unwanted changes and seamless data capture
  • Large LCD with bright white backlight enables viewing in dim light
  • Avoid neck strain—tilting display easily adjusts to different viewing angles
The HV-C/CP and HW-C/CP bench scales feature a bright 3-5-7 segment color (red-green-yellow) comparator function for checkweighing. A 10-digit keypad makes entering comparator limits and tare weights quick and easy, while a key lock function prevents operators from making unnecessary setting changes. The negative checkweighing function allows you to easily move parts from a master carton on the scale to containers on the production line.
Weighing modes include parts counting and accumulation function for totaling weigh data. Weighing units include kg, lb, and oz.
$1,753.33 - $3,410.90CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
A&D Weighing HV-15KC Bench Scale, 3/6/15 kg x 0.001/0.002/0.005 kg; NTEP Approved 1112450 Mfr # HV-15KC
COMPARE
15 30 Bench Scale, 3/6/15 kg x 0.001/0.002/0.005 kg; NTEP Approved
$1,753.33CAD / Each
A&D Weighing HV-60KC Bench Scale, 15/30/60 kg x 0.005/0.01/0.02 kg; NTEP Approved 1112452 Mfr # HV-60KC
COMPARE
60 132 Bench Scale, 15/30/60 kg x 0.005/0.01/0.02 kg; NTEP Approved
$1,753.33CAD / Each
A&D Weighing HV-200KC Bench Scale, 60/150/220 kg x 0.02/0.05/0.1 kg; NTEP Approved 1112454 Mfr # HV-200KC
COMPARE
220 500 Bench Scale, 60/150/220 kg x 0.02/0.05/0.1 kg; NTEP Approved
$2,210.09CAD / Each
A&D Weighing HW-10KC High Resolution Bench Scale; 10 kg x 0.001 kg 1112456 Mfr # HW-10KC
COMPARE
10 20 High Resolution Bench Scale; 10 kg x 0.001 kg
$1,922.77CAD / Each
A&D Weighing HW-10KCP High Resolution Bench Scale with Printer; 10 kg x 0.001 kg 1112457 Mfr # HW-10KCP
COMPARE
10 20 High Resolution Bench Scale with Printer; 10 kg x 0.001 kg
$2,909.95CAD / Each
A&D Weighing HW-60KC High Resolution Bench Scale; 60 kg x 0.005 kg 1112458 Mfr # HW-60KC
COMPARE
60 150 High Resolution Bench Scale; 60 kg x 0.005 kg
$2,070.11CAD / Each
A&D Weighing HW-60KCP High Resolution Bench Scale with Printer; 60 kg x 0.005 kg 1112459 Mfr # HW-60KCP
COMPARE
60 150 High Resolution Bench Scale with Printer; 60 kg x 0.005 kg
$3,116.22CAD / Each
A&D Weighing HW-100KC High Resolution Bench Scale; 100 kg x 0.01 kg 1112460 Mfr # HW-100KC
COMPARE
100 200 High Resolution Bench Scale; 100 kg x 0.01 kg
$2,173.25CAD / Each
A&D Weighing HW-100KCP High Resolution Bench Scale with Printer; 100 kg x 0.01 kg 1112461 Mfr # HW-100KCP
COMPARE
100 200 High Resolution Bench Scale with Printer; 100 kg x 0.01 kg
$3,293.03CAD / Each
A&D Weighing HW-200KC High Resolution Bench Scale; 220 kg x 0.02 kg 1112462 Mfr # HW-200KC
COMPARE
220 500 High Resolution Bench Scale; 220 kg x 0.02 kg
$2,335.32CAD / Each
A&D Weighing HW-200KCP High Resolution Bench Scale with Printer; 220 kg x 0.02 kg 1112463 Mfr # HW-200KCP
COMPARE
220 500 High Resolution Bench Scale with Printer; 220 kg x 0.02 kg
$3,410.90CAD / Each
Showing 1- 11 of 11