A&D Weighing HD Counting Scales
Large wide angle LCD display with detachable display head for use with large sized containers and for flexible reading and operation
  • Designed for cost-conscious users
  • High quality, full featured digital counting scales
  • Automatic Counting Accuracy Improvement (ACAI) for excellent counting accuracy
  • New electronic design for improved operation
  • •Comparator function for packaging or check weighing operations
  • M+ totaling for running totals
  • AC power, optional dry cell battery or optional rechargeable battery operation
  • Detachable display head
  • Large weighing pan for oversize containers
  • Automatic power off saves battery life
  • Intuitive operation for ease of use and reduced operator training.
The HD Series high capacity electronic counting balances provide high performance and features at a low price. These portable digital counting scales include a detachable display and large weighing pan for oversize containers. •Choice of simple operation (A version) or 10 button keypad models for direct entry of tare weight, piece weight & sample size (B version)
$1,989.08 - $2,232.19CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing HD-12KA Counting Scale 30lb x 0.005 Lb / 12kg x 2g 1122975 Mfr # HD-12KA
COMPARE
12 30 115
$1,989.08CAD / Each
A&D Weighing HD-30KA Counting Scale 60lb x 0.01 Lb / 30kg x 5g 1122976 Mfr # HD-30KA
COMPARE
30 60 115
$1,989.08CAD / Each
A&D Weighing HD-60KA Counting Scale 120lb x 0.02 Lb / 60kg x 10g 1122977 Mfr # HD-60KA
COMPARE
60 120 115
$1,989.08CAD / Each
A&D Weighing HD-12KB Counting Scale 30lb x 0.005 Lb / 12kg x 2g 1122978 Mfr # HD-12KB
COMPARE
12 30 115
$2,232.19CAD / Each
A&D Weighing HD-30KB Counting Scale 60lb x 0.01 Lb / 30kg x 5g 1122979 Mfr # HD-30KB
COMPARE
30 60 115
$2,210.09CAD / Each
A&D Weighing HD-60KB Counting Scale 120lb x 0.02 Lb / 60kg x 10g 1122980 Mfr # HD-60KB
COMPARE
60 120 115
$2,210.09CAD / Each