A&D Weighing Density Determination Kits
Use your balance to determine density with these convenient kits
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.
$103.14 - $1,583.89CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
A&D Weighing GXA-13 Density Determination Kit (For The 1 Mg Models Only) 1110979 Mfr # GXA-13
COMPARE
Density Determination Kit (For The 1 Mg Models Only)
$979.80CAD / Each
A&D Weighing AD-1653 Density Determination Kit for BM, GH, HR-I, GR, HR, and HA Series Balances 1111120 Mfr # AD-1653
COMPARE
Density Determination Kit for Hr-A/Hr-Az
$1,119.78CAD / Each
A&D Weighing AD-1653 Density Determination Kit for BA, BM, GH, HR-I, GR, HR, and HA Series Balances 1111474 Mfr # AD-1653
COMPARE
Density Determination Kit ,
$1,119.78CAD / Each
A&D Weighing AD-1654 Density Determination Kit for HR-AZ, HR-A, FZ- I, and, FX- i Series Balances 1111475 Mfr # AD-1654
COMPARE
Density Determination Kit ,
$1,583.89CAD / Each
A&D Weighing EJ-13 Density Determination Kit for EJ Series Compact Precision Balance 1111507 Mfr # EJ-13
COMPARE
Density Determination Kit ,
$162.07CAD / Each
A&D Weighing EKW-07I Under Hook for EK-i/EW-i Series Magnetic Material and Density Measurements 1111992 Mfr # EKW-07I
COMPARE
Under Hook for Magnetic Material and Density Measurements
$103.14CAD / Each