A&D Weighing EKW-07I Under Hook for EK-i/EW-i Series Magnetic Material and Density Measurements
Representative image only.
A&D Weighing EKW-07I Under Hook for EK-i/EW-i Series Magnetic Material and Density Measurements
A&D Weighing | Mfr: EKW-07I| Item: 1111992
Use your balance to determine density with these convenient kits
$103.14CAD / Each
$103.14CAD / Each
Buy It Together
Current Item
A&D Weighing EKW-07I Under Hook for EK-i/EW-i Series Magnetic Material and Density Measurements
A&D Weighing EKW-07I Under Hook for EK-i/EW-i Series Magnetic Material and Density Measurements
1111992
$103.14CAD / Each
+
Cole-Parmer RS-232 to USB Cable
1010083
$76.32CAD / Each
=
Total Price 179.46 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand the capabilities of your balance with these convenient kits
  • Ensure the kit corresponds to your correct A&D Weighing balance
  • 1 year warranty
More About this Item
These density determination kits, when combined with one of the A&D Weighing electronic balances, allows the density of a solid and liquid to be measured easily. Balances sold separately.
1 Year Warranty
Specifications
  • DescriptionUnder Hook for Magnetic Material and Density Measurements