A&D Weighing AD-4212C Production Weighing Sensor
High-precision weighing and adjustable speed to stabilization optimize production line weighing
  • FAST, MID, and SLOW stabilization settings let you choose the right combination of response time and sensitivity for your application
  • Compact footprint and mounting fixtures make integration into existing production line systems easy
  • Create your own weighing pan to better suit your samples
  • Separate sensor and display units are perfect for isolated weighing — such as inside vacuum chambers, fume hoods and glove boxes
  • Display unit can be panel or wall mounted
  • Patented shock absorber fitted under the weighing pan protects sensor from vertical and transverse overloads
  • Standard bidirectional RS-232 interface connects to PC for data download and control
  • Output conforms to GLP/GMP requirements
The extremely quick Super Hybrid Sensor technology makes this balance ideal for your high-speed weighing and filling, checkweighing, and remote weighing applications. Response times are as low as 0.5 seconds for weighing in a fast-paced environment or filling operations. The compact weighing sensor can be placed anywhere in a production line and allows for a custom-designed weighing pan. The AD-4212 series of weighing sensors provides GLP/GMP compliance with data output of weight and calibration data, as well as unit mass information for the counting mode or over/under limit values.
The data memory function provides storage of 200 weight measurements, 100 weight measurements with time and date, 50 calibration and calibration test measurements, 20 unit weight measurements for counting mode or 20 comparator settiing limits values.
The AD-4212C is rated IP65 to withstand dust or powder spills, and when washdown is necessary.
$7,308.01 - $11,028.32CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
A&D Weighing AD-4212C-300 Precision Sensor with IP-65 SS Housing, 320g x 1mg 1111608 Mfr # AD-4212C-300
COMPARE
320 115
$7,308.01CAD / Each
A&D Weighing AD-4212C-301 Precision Sensor IP-65 SS Housing, 51/320g x 0.1/1mg 1111609 Mfr # AD-4212C-301
COMPARE
320 115
$11,028.32CAD / Each
A&D Weighing AD-4212C-600 Precision Sensor with IP-65 SS Housing, 620g x 1mg 1111611 Mfr # AD-4212C-600
COMPARE
620 115
$11,028.32CAD / Each
A&D Weighing AD-4212C-3000 Precision Sensor with IP-65 SS Housing, 3200g x 10mg 1111612 Mfr # AD-4212C-3000
COMPARE
3200 115
$7,308.01CAD / Each
A&D Weighing AD-4212C-3100 Precision Sensor IP-65 SS Housing, 510/3200g x 1/10mg 1111613 Mfr # AD-4212C-3100
COMPARE
3200 115
$11,028.32CAD / Each
A&D Weighing AD-4212C-6000 Precision Sensor with IP-65 SS Housing, 6200g x 1mg 1111614 Mfr # AD-4212C-6000
COMPARE
6200 115
$11,028.32CAD / Each