A&D Newton EJ Portable Balances
Get flexibility and speed—delivers fast one-second response time
  • Easy-to-read LCD with backlight provides clear view of results
  • Specific gravity, percentage weighing, and parts counting functions
  • Comparator function gives a visual reference against a set target weight
  • Flexible—AC or battery operation
  • Feature low-battery, overload and stability indicators
  • Customize your balance using the configurable key mode
  • Stackable body allows convenient storage of multiple units
These lightweight and portable balances offer quick five-button operation -- on/off, sample, zero/tare, print, and change units. The stainless steel weigh pan and ABS plastic housing make for easy cleaning. The backlight clear LCD with 16-mm high characters can be read easily in any light condition and from various angles. Other features include stability icon, front level bubble, four leveling feet, security slot for lock down capability, and display hold.
Optional RS-232 and USB interface plug-ins are available to meet your communication needs.
$515.69 - $1,245.01CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
A&D Weighing EJ-120 Newton Portable Balance, 120g x 0.01g; 120V 1110961 Mfr # EJ-120
COMPARE
External 120 0.01
$515.69CAD / Each
A&D Weighing EJ-200 Newton Portable Balance, 210g x 0.01g; 120V 1110962 Mfr # EJ-200
COMPARE
External 210 0.01
$640.92CAD / Each
A&D Weighing EJ-300 Newton Portable Balance, 310g x 0.01g; 120V 1110963 Mfr # EJ-300
COMPARE
External 310 0.01
$766.16CAD / Each
A&D Weighing EJ-410 Newton Portable Balance, 410g x 0.01g; 120V 1110964 Mfr # EJ-410
COMPARE
External 410 0.01
$825.10CAD / Each
A&D Weighing EJ-610 Newton Portable Balance, 610g x 0.01g; 120V 1110965 Mfr # EJ-610
COMPARE
External 610 0.01
$994.54CAD / Each
A&D Weighing EJ-1500 Newton Portable Balance, 1500g x 0.1g; 120V 1110966 Mfr # EJ-1500
COMPARE
External 1500 0.1
$515.69CAD / Each
A&D Weighing EJ-2000 Newton Portable Balance, 2100g x 0.1g; 120V 1110967 Mfr # EJ-2000
COMPARE
External 2100 0.1
$626.19CAD / Each
A&D Weighing EJ-3000 Newton Portable Balance, 3100g x 0.1g; 120V 1110968 Mfr # EJ-3000
COMPARE
External 3100 0.1
$773.53CAD / Each
A&D Weighing EJ-4100 Newton Portable Balance, 4100g x 0.1g; 120V 1110969 Mfr # EJ-4100
COMPARE
External 4100 0.1
$861.93CAD / Each
A&D Weighing EJ-6100 Newton Portable Balance, 6100g x 0.1g; 120V 1110970 Mfr # EJ-6100
COMPARE
External 6100 0.1
$979.80CAD / Each
A&D Weighing EJ-1202 Newton Portable Balance, 1200g x 0.01g; 120V 1110975 Mfr # EJ-1202
COMPARE
External 1200 0.01
$1,090.31CAD / Each
A&D Weighing EJ-3002 Newton Portable Balance, 3100g x 0.01g; 120V 1110976 Mfr # EJ-3002
COMPARE
External 3100 0.01
$1,171.35CAD / Each
A&D Weighing EJ-54D2 Newton Portable Balance with Breeze Break, 22/52g x 0.2/1mg; 120V 1111352 Mfr # EJ-54D2
COMPARE
External 52 0.001
$1,245.01CAD / Each
A&D Weighing EJ-123 Newton Portable Balance with Breeze Break, 120g x 0.001g; 120V 1111596 Mfr # EJ-123
COMPARE
External 120 0.001
$876.67CAD / Each
A&D Weighing EJ-303 Newton Portable Balance with Breeze Break, 310g x 0.001g; 120V 1111597 Mfr # EJ-303
COMPARE
External 310 0.001
$1,046.11CAD / Each
Showing 1- 15 of 15