Whatman 6703-7511 Disposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb
Representative image only.
Whatman 6703-7511 Disposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb
Whatman | Mfr: 6703-7511| Item: 2970142
Use polypropylene (PP) membrane filters for solvents, clean air/gas, prefiltration, and sample preparation
$172.90CAD / Pkg of 1
$172.90CAD / Pkg of 1
Buy It Together
Current Item
Whatman 6703-7511 Disposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb
Whatman 6703-7511 Disposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb
2970142
$172.90CAD / Pkg of 1
+
Whatman 6702-3600 HEPA-CAP Filter; 36 capsule, 1pk
2970092
$175.26CAD / Pkg of 1
=
Total Price 348.16 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Autoclavable
  • 45 day warranty
More About this Item
Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
Specifications
  • Membrane MaterialPolypropylene
  • Pore Size (µm)10
  • Effective Filter Area (cm²)400
  • Connections1/2" Barb
  • DescriptionDisposable Capsule Filters, PP membrane, 10um, 1/2" stepped barb
  • Warranty45 days