Welch CRVpro8 Rotary Vane Vacuum Pump, 5.6 CFM, XPRF 115V/208-230V 60 Hz

Representative image only.

Welch CRVpro8 Rotary Vane Vacuum Pump, 5.6 CFM, XPRF 115V/208-230V 60 Hz

Welch | Mfr: 3086E-01| Item: 7920621

Ideal for hazardous areas where flammable gases or vapors are present

$8,914.01CAD / Each

$8,914.01CAD / Each

Buy It Together

Current Item

Welch CRVpro8 Rotary Vane Vacuum Pump, 5.6 CFM, XPRF 115V/208-230V 60 Hz
Welch CRVpro8 Rotary Vane Vacuum Pump, 5.6 CFM, XPRF 115V/208-230V 60 Hz
7920621

$8,914.01CAD / Each
+ +
Welch Exhaust Filter For CRVpro 2, 4, 6, 8, NW16
7920623

$804.47CAD / Each
=
Total Price 14669.07 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Directorr™ Premium vacuum pump oil, centering rings, and clamping rings.

Key Features

  • Consumes less oil by cool running operation
  • Large oil capacity minimizes rates of oil breakdown
  • Chemical resistant coating in oil case and pump module protects non-moving parts
  • Up to 10 dB noise level for undisturbed work
  • IP54 enclosure rating
  • Flow rate of 5.6 CFM (158 l/min)
  • 1 year warranty

More About this Item

The CRVpro series two-stage, direct drive rotary vane vacuum pumps are fitted with Class 1, Groups C and D, and Class 2, Groups F and G explosion proof motors. Be sure to use a certified electrician for proper installation of these pumps. Conduit wiring installation required. No cord, plug, or switch provided.

A unique air flow design allows the pumps to run 10°C cooler than standard rotary vane pumps, leading to reduced chemical activity within the pump and less oil consumption—up to 36% less than comparable pumps. The oil-sealed pumps feature a gas ballast, forced oil lubrication, anti-suck back valve, low noise level, and thermal overload protection. The gas ballast uses either outside air or dry inert gas to reduce condensable vapors from entering the pump oil.

Ideal for use in freeze drying, degassing, vacuum ovens, and concentrators.

Conformite EuropeenneCanadian Standards AssociationIP54 Rated1 Year Warranty
Specifications
  • Wetted PartsPTFE
  • Free Air Capacity (CFM)5.6
  • Free Air Capacity (LPM)158
  • Max Vacuum (torr)0.05
  • Max Vacuum (with ballast)7x10-2 mbar (5x10-2 Torr)
  • Max Vacuum (without ballast)2x10-3 mbar (1.5x10-3 Torr)
  • Outlet PortNW 16 flange
  • Noise Level≤50 dB (A)
  • Oil Capacity (Liters)1
  • Oil Capacity (oz)34
  • Oil Capacity (mL)1000
  • Power (VAC)115
  • Power (Hz)60
  • Height (in)9
  • Width (in)6.2
  • Length (in)18.2
  • Height (cm)23
  • Width (cm)15.7
  • Length (cm)46.3
  • Motor hp1/2
  • DescriptionCRVpro8 Rotary Vane Vacuum Pump, 5.6 CFM, XPRF 115V/208-230V 60 Hz
  • Warranty1 Year
Reviews
Q&A