Traceable Mini Digital Benchtop Atomic Clock

Representative image only.

Traceable Mini Digital Benchtop Atomic Clock

Traceable | Mfr: 1065TR| Item: 6200208

Extremely accurate—to 1/10,000 of a second!

$48.92CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$48.92CAD / Each

Buy It Together

Current Item

Traceable Mini Digital Benchtop Atomic Clock
Traceable Mini Digital Benchtop Atomic Clock
6200208

$48.92CAD / Each
+
Traceable Monitoring Hygrometer
2025030

$91.20CAD / Each
+ =
Total Price 312.87 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Traceable® Certificate and batteries.

Key Features

  • Clear display with elegant acrylic lens shows a resolution of 1 second
  • Displays time, calendar, and weekday
  • Reads indoor temperature (°C/°F)
  • Offers alarm and snooze function
  • Use an amber LED backlight for daytime use and an auto-night light function for nighttime
  • Mini size—only measures 2.5"W x 2.5"H
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

These digital wall clocks contain a pre-tuned internal radio receiver that receives a radio frequency signal generated by the U.S. Atomic Clock in Boulder, Colorado. With an accuracy of 1/10,000 of a second, the clocks automatically adjust to precise time (12 or 24-hr format), daylight savings time, month, date, and day of the week. Also view the ambient temperature, signal strength, and time zone map on the LCD.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

NIST Traceable Calibration1 Year Warranty
Specifications
  • Clock12 or 24-hr format
  • Resolution1 sec
  • Accuracy±0.0001 sec
  • AlarmYes
  • DisplayLCD
  • CertificationsNIST-traceable
  • Temperature Resolution0.1°
  • Max Temperature (° F)122
  • Min Temperature (° F)23
  • Max Temperature (° C)50
  • Min Temperature (° C)-5
  • Battery Type2 AAA (included)
  • Height (InchShort)2 1/2
  • Depth (in)1
  • Width (InchShort)2 1/2
  • Height (cm)6.35
  • Depth (cm)2.54
  • Width (cm)6.35
  • DescriptionMini Digital Benchtop Atomic Clock
  • Warranty1 year
Reviews
Q&A