Traceable® Certificate, wall-mount screw, and batteries.
Key Features
Displays month, date, day of the week, and time!
Increase versatility with alarm and snooze options
Receives a 60 kHz frequency signal generated by the U.S. Atomic Clock in Boulder, Colorado
Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
1 year warranty
More About this Item
These digital wall clocks contain a pre-tuned internal radio receiver that receives a radio frequency signal generated by an atomic clock. With an accuracy of 1/10,000 of a second, the clocks automatically adjust to precise time, leap year, and leap seconds (earth rotation wobble corrections). View month, date, day of the week, and time (hours, minutes, and seconds) in a.m./p.m. or 24-hour time as well as ambient temperature and relative humidity. The LCD features digits as large as 3-5/8”H for visibility from up to 20 feet away.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Automatically adjusts clock to daylight savings time.
Guaranteed to have at least one year of calibration remaining on certificate.
Specifications
Timing Capacity24 hours
Count DirectionUp
Clock12 or 24-hour format
Resolution1 sec
Accuracy±0.0001 sec
DisplayLCD, 3-5/8" H
CertificationsNIST-traceable
Temperature Resolution0.1°F/C
Max Temperature (° F)113
Min Temperature (° F)32
Max Temperature (° C)45
Min Temperature (° C)0
Battery Type2 AA (included)
Height (InchShort)8 3/4
Depth (in)7/8
Width (InchShort)8 3/4
Height (cm)22.2
Depth (cm)2.2
Width (cm)22.2
Signal TypeUS
Mounting TypeWall
DescriptionRadio-Controlled Digital Wall Clock, US Radio Signal