Thermo Scientific P10DS-1 Standard Double-Sided Vertical Cell, 20 x 20 cm gel size

Representative image only.

Thermo Scientific P10DS-1 Standard Double-Sided Vertical Cell, 20 x 20 cm gel size

Thermo Scientific | Mfr: P10DS-1| Item: 2855415

Larger-size cassette gives you the maximum number of samples and separation length

$3,801.35CAD / Each

$3,801.35CAD / Each

Buy It Together

Current Item

Thermo Scientific P10DS-1 Standard Double-Sided Vertical Cell, 20 x 20 cm gel size
Thermo Scientific P10DS-1 Standard Double-Sided Vertical Cell, 20 x 20 cm gel size
2855415

$3,801.35CAD / Each
+
Enduro E0160 Gel XL Electrophoresis System 120V
1731411

$1,735.65CAD / Each
+ =
Total Price 5627.02 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

upper buffer chamber with internal cooling core, lower buffer chamber, safety lid with attached power supply leads, four blank glass plates, four notched glass plates, blocking plate for single gel operation, and 1.5-mm thick spacer set.

Key Features

  • Upper buffer chamber is extended, providing quick and easy gel casts
  • Built-in cooling system circulates cold tap water for fast run times and flat, even banding patterns
  • Simple clamping mechanism for trouble-free loading of glass cassettes and precast gels
  • 3 year warranty

More About this Item


Model 28554-15 also includes two 1.5-mm thick 15-well combs and two 1.5-mm thick 20-well combs.

3 Year Warranty
Specifications
  • Buffer Solution Required (mL)1400
  • Gel Thickness (mm)1.5
  • Cell Width (in)6.5
  • Cell Height (in)9.5
  • Cell Depth (in)12
  • Cell Width (cm)16.51
  • Cell Height (cm)24.13
  • Cell Depth (cm)30.48
  • Glass Width (in)7.87
  • Glass Length (in)7.87
  • Glass Width (cm)20
  • Glass Length (cm)20
  • DescriptionStandard Double-Sided Vertical Cell, 20 x 20 cm gel size
  • Warranty3 year
Reviews
Q&A