Thermo Scientific Dual Star pH/ISE/mV Meter with Electrode

Representative image only.

Thermo Scientific Dual Star pH/ISE/mV Meter with Electrode

Thermo Scientific | Mfr: 2115101| Item: 5882510

Innovative EZ Startup™ ensures no manual is needed to begin taking measurements

$4,667.48CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$4,667.48CAD / Each

Buy It Together

Current Item

Thermo Scientific Dual Star pH/ISE/mV Meter with Electrode
Thermo Scientific Dual Star pH/ISE/mV Meter with Electrode
5882510

$4,667.48CAD / Each
+ + =
Total Price 6874.98 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Dual inputs allow you to simultaneously measure and display pH, ISE, and temperature
  • Simple to operate — prompts walk you through meter operation and setup
  • 3 year warranty

More About this Item

The Orion Dual Star meter provides dual BNC and temperature inputs for simultaneous pH and ISE readings. Ion determination is facilitated by additional options for concentration units, optional auto-blank for low-level measurements, built-in Star-stirrer control, and the ability to view, print your calibration graph with slope data, and modify calibration points. The meter easily guides you through common direct measurements, but also is programmed for a range of single and double known addition and subtraction methods. Autosampler capabilities allow you to take advantage of high sample throughput technology to save time and improve measurement quality.

The Dual Star utilizes EZ Startup™ for setup without the manual, and Smart Stability™ and SmartAveraging™ to ensure accurate results. The meter offers a full complement of GLP-related features including 1000 point datalogging with time/date stamp, electrode verification, and full calibration data. Generate and store up to 10 password-protected methods per channel. Easily download from the USB port or bidirectional RS-232.

Meter includes: Quick-Start guide, electrode stand, stainless steel ATC probe, and universal AC power supply.

Meter with electrode adds: a refillable, glass-body Orion ROSS Ultra® electrode.

Meter kit adds: 475-mL bottles of 4, 7, and 10 buffer; and Orion ROSS® electrode storage solution.

UL Listed.Canadian Standards AssociationIP54 Rated3 Year WarrantyISO9001:2000 Certified Supplier
Specifications
  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.2% mV or 0.05% whichever is greater
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)90 to 260
  • Width (cm)22.86
  • Height (cm)5.08
  • Width (in)9
  • Length (in)8
  • Height (in)2
  • DescriptionpH/ISE/mV Meter with Electrode
  • Warranty3 year
Reviews
Q&A