Thermo Scientific 900063 Optimum Results D Filling Solution for ISE; 5 x 60mL Bottles
Representative image only.
Thermo Scientific 900063 Optimum Results D Filling Solution for ISE; 5 x 60mL Bottles
Thermo Scientific | Mfr: 900063| Item: 0571336
Optimize water analysis
$205.02CAD / Each
$205.02CAD / Each
Buy It Together
Current Item
Thermo Scientific 900063 Optimum Results D Filling Solution for ISE; 5 x 60mL Bottles
Thermo Scientific 900063 Optimum Results D Filling Solution for ISE; 5 x 60mL Bottles
0571336
$205.02CAD / Each
+ =
Total Price 1523.11 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Extend the life of your ion selective electrodes with filling solutions
More About this Item
Specially formulated ISE filling solutions ensure minimal electrode interferences to provide accurate measurements.
Fill solution for Bromide, Cupric and Iodide Iion-selective electrodes (ISE).
Specifications
  • Capacity (mL)5 x 60
  • Compatible WithOrion Ion Selective Electrodes
  • DescriptionOptimum Results D Filling Solution for ISE; 5 x 60mL Bottles