Testo 405i Smart Probe Hot-Wire Anemometer for use with Smartphone or Tablet
Representative image only.
Testo 405i Smart Probe Hot-Wire Anemometer for use with Smartphone or Tablet
Testo | Mfr: 0560 1405 01| Item: 3000919
View HVAC measurement on your Smart device from up to 60 feet away!
$307.94CAD / Each
for an additional
$529.68CAD / Each
for an additional
$660.82CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$307.94CAD / Each
Buy It Together
Current Item
Testo 405i Smart Probe Hot-Wire Anemometer for use with Smartphone or Tablet
Testo 405i Smart Probe Hot-Wire Anemometer for use with Smartphone or Tablet
3000919
$307.94CAD / Each
+ + +
Testo Smart Probe VAC Case
3000948
$88.40CAD / Each
=
Total Price 1274.48 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Three AAA batteries and certificate of calibration.
Key Features
  • Use the Smart Probes app to read and create custom reports
  • Save and share measurements as a PDF or Excel® file
  • Perform difficult volume flow calculations automatically
  • Multi-color LED displays low battery warning and Bluetooth® connectivity
  • 2 year warranty
More About this Item
This compact, rugged, easy-to-use wireless hot-wire anemometer offers accurate measurements of air velocity, volume flow, and temperature in ventilation ducts with the use of a 15” telescopic shaft – ideal for the HVAC professional.
Easily monitor and transmit all measured data wirelessly via Bluetooth® to your smartphone or tablet from up to 60 feet away. With the free Testo Smart Probes app, you can display measurements from up to six Smart Probes at the same time, visualize measurement value changes as a graph or a table, and perform automatic calculations. Easily turn your data into a custom report, attach images created with the app, and send via email as a PDF or Excel® file.
Combine this probe with the 410i Smart Probe vane anemometer, 605i Smart Probe thermohygrometer, and 805i Smart Probe infrared thermometer to monitor HVAC-R applications.
Available for sale in the United States and Canada only.
Two year warrantyApple App StoreConformite EuropeenneGoogle Play App
Specifications
  • Max Air Velocity (ft/min)0
  • Min Air Velocity (ft/min)6000
  • Air Velocity Resolution2 fpm
  • Air Velocity Accuracy±(9.9 fpm + 2.5 % of m.v.) (0 to 400 fpm), ±(29.5 fpm + 2.5 % of m.v.) (400 to 3000 fpm)
  • Min Temperature (° F)-4
  • Max Temperature (° F)140
  • Min Temperature (° C)-2
  • Max Temperature (° C)60
  • Temperature Accuracy ±0.9°F
  • Temperature Resolution0.1°
  • Battery3 AAA (included)
  • DescriptionSmart Probe Hot-Wire Anemometer