SHOWA 878-08 Gloves, heavy-weight, medium
Representative image only.
SHOWA 878-08 Gloves, heavy-weight, medium
SHOWA | Mfr: 878-08| Item: 8635720
Perfect for chemical handling, spill control, and hazmat applications
$166.12CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$166.12 CAD / Pair of 2
  • 10+$149.51 CAD / Pair of 2
$166.12CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$166.12 CAD / Pair of 2
  • 10+$149.51 CAD / Pair of 2
Buy It Together
Current Item
SHOWA 878-08 Gloves, heavy-weight, medium
SHOWA 878-08 Gloves, heavy-weight, medium
8635720
$166.12CAD / Pair of 2
+ =
Total Price 564.67 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • Thickness (mil)30
  • MaterialButyl
  • SizeMedium
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripSmooth
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, heavy-weight, medium