SHOWA 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair
Representative image only.
SHOWA 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair
SHOWA | Mfr: 892-10| Item: 8628823
Get flexible, heavy-duty protection
$398.55CAD / Pair of 1
BUY MORE & SAVE
  • 1-11$398.55 CAD / Pair of 1
  • 12+$358.70 CAD / Pair of 1
$398.55CAD / Pair of 1
BUY MORE & SAVE
  • 1-11$398.55 CAD / Pair of 1
  • 12+$358.70 CAD / Pair of 1
Buy It Together
Current Item
SHOWA 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair
SHOWA 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair
8628823
$398.55CAD / Pair of 1
+
SHOWA 874-08 Gloves, smooth, medium
8635701
$148.44CAD / Pair of 1
=
Total Price 546.99 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Superior resistance to PCBs
More About this Item
Made of Viton® fluoroelastomer, these gloves provide superior resistance to PCBs, chlorinated and aromatic solvents, and vapors. Smooth, flexible, unlined gloves with a rolled cuff are injection-molded for consistent thickness throughout.
Conformite Europeenne
Specifications
  • Thickness (mil)12
  • MaterialViton
  • SizeX-Large
  • Length (in)12
  • Length (cm)30.5
  • ColorBlack
  • SterileNo
  • DescriptionViton Gloves, extra-large, 12-mil thickness, 1 pair