SHOWA 874R-9 Gloves, textured, large
Representative image only.
SHOWA 874R-9 Gloves, textured, large
SHOWA | Mfr: 874R-9| Item: 8635712
Perfect for chemical handling, spill control, and hazmat applications
$123.91CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$123.91 CAD / Pair of 2
  • 10+$111.52 CAD / Pair of 2
$123.91CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$123.91 CAD / Pair of 2
  • 10+$111.52 CAD / Pair of 2
Buy It Together
Current Item
SHOWA 874R-9 Gloves, textured, large
SHOWA 874R-9 Gloves, textured, large
8635712
$123.91CAD / Pair of 2
+ =
Total Price 522.46 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • Thickness (mil)14
  • MaterialButyl
  • SizeLarge
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripTextured
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, textured, large