SHOWA 874R-8 Gloves, textured, medium
Representative image only.
SHOWA 874R-8 Gloves, textured, medium
SHOWA | Mfr: 874R-8| Item: 8635711
Perfect for chemical handling, spill control, and hazmat applications
$86.78CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$86.78 CAD / Pair of 1
  • 10+$78.10 CAD / Pair of 1
$86.78CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$86.78 CAD / Pair of 1
  • 10+$78.10 CAD / Pair of 1
Buy It Together
Current Item
SHOWA 874R-8 Gloves, textured, medium
SHOWA 874R-8 Gloves, textured, medium
8635711
$86.78CAD / Pair of 1
+ =
Total Price 485.33 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Conformite Europeenne
Specifications
  • MaterialButyl
  • Thickness (mil)14
  • SizeMedium
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripTextured
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, textured, medium