SHOWA 874R-10 Gloves, textured, extra-large
Representative image only.
SHOWA 874R-10 Gloves, textured, extra-large
SHOWA | Mfr: 874R-10| Item: 8635713
Perfect for chemical handling, spill control, and hazmat applications
$126.86CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$126.86 CAD / Pair of 2
  • 10+$114.17 CAD / Pair of 2
$126.86CAD / Pair of 2
BUY MORE & SAVE
  • 1-9$126.86 CAD / Pair of 2
  • 10+$114.17 CAD / Pair of 2
Buy It Together
Current Item
SHOWA 874R-10 Gloves, textured, extra-large
SHOWA 874R-10 Gloves, textured, extra-large
8635713
$126.86CAD / Pair of 2
+ =
Total Price 525.41 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • MaterialButyl
  • Thickness (mil)14
  • SizeX-Large
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripTextured
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, textured, extra-large