SHOWA 874-10 Gloves, smooth, extra-large
Representative image only.
SHOWA 874-10 Gloves, smooth, extra-large
SHOWA | Mfr: 874-10| Item: 8635704
Perfect for chemical handling, spill control, and hazmat applications
$101.52CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$101.52 CAD / Pair of 1
  • 10+$91.37 CAD / Pair of 1
$101.52CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$101.52 CAD / Pair of 1
  • 10+$91.37 CAD / Pair of 1
Buy It Together
Current Item
SHOWA 874-10 Gloves, smooth, extra-large
SHOWA 874-10 Gloves, smooth, extra-large
8635704
$101.52CAD / Pair of 1
+ =
Total Price 500.07 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • MaterialButyl
  • Thickness (mil)14
  • SizeX-Large
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripSmooth
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, smooth, extra-large