SHOWA 874-09 Gloves, smooth, 14 mil, large
Representative image only.
SHOWA 874-09 Gloves, smooth, 14 mil, large
SHOWA | Mfr: 874-09| Item: 8635702
Perfect for chemical handling, spill control, and hazmat applications
$148.44CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$148.44 CAD / Pair of 1
  • 10+$133.60 CAD / Pair of 1
$148.44CAD / Pair of 1
BUY MORE & SAVE
  • 1-9$148.44 CAD / Pair of 1
  • 10+$133.60 CAD / Pair of 1
Buy It Together
Current Item
SHOWA 874-09 Gloves, smooth, 14 mil, large
SHOWA 874-09 Gloves, smooth, 14 mil, large
8635702
$148.44CAD / Pair of 1
+ =
Total Price 546.99 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
These gloves provide excellent chemical resistance to hazardous materials — gases, aldehydes, amines, and ketones — without being bulky or restrictive. They are unlined to ensure an excellent grip. Gloves are 14" long with rolled cuffs that fit all hazmat suits.
Compliance: ASTM F-739
Specifications
  • MaterialNitrile
  • Thickness (mil)14
  • SizeLarge
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • GripSmooth
  • CuffRolled
  • ComplianceASTM F739
  • SterileNo
  • DescriptionGloves, smooth, 14 mil, large