Secador 5.0 Auto-Desiccator Cabinet

Representative image only.

Secador 5.0 Auto-Desiccator Cabinet

Secador | Mfr: 420571003| Item: 0500629

Reliable dry storage for humidity sensitive products

$3,698.21CAD / Each

$3,698.21CAD / Each

Buy It Together

Current Item

Secador 5.0 Auto-Desiccator Cabinet
Secador 5.0 Auto-Desiccator Cabinet
0500629

$3,698.21CAD / Each
+
Secador 5.0 Desiccator Shelf
0500628

$122.00CAD / Each
+ =
Total Price 3820.21 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Large clear polycarbonate panels
  • LCD displays internal temperature and humidity level
  • Three adjustable shelves
  • 1 year warranty

More About this Item

These lightweight desiccator cabinets offer dry storage for humidity sensitive products such as laboratory reagents and samples, optical equipment, electronics, and 3D printing filament. The design features high density foam gaskets which keep ambient humidity out and dry air inside. Clear polycarbonate panels allow you to view contents within the cabinets, while the LCD panel located on the outside for the units clearly show the internal temperature and humidity level. Three removable shelves can easily be adjusted within the 1.6 cu ft cabinet interior to meet various height requirements of your products. Additionally shelves can be purchased separately (05006-28).

Two models are available. The electronic model allows you to operate the unit from a 100 to 240 VAC power source without the need of a converter and keeps internal humidity at 10-40%. The non-electronic model can be used with any other Bel-Art desiccant product.

1 Year Warranty
Specifications
  • Chamber Volume (cu ft)1.6
  • Chamber Volume (Liters)47.6
  • Interior Width (in)11
  • Interior Height (in)19.685
  • Interior Depth (in)13.385
  • Interior Width (cm)28
  • Interior Height (cm)50
  • Interior Depth (cm)34
  • Exterior Width (in)11.8
  • Exterior Height (InchShort)25
  • Exterior Depth (in)15
  • Exterior Width (cm)30
  • Exterior Height (cm)63.5
  • Exterior Depth (cm)38
  • MaterialPolycarbonate panels & shelves, ABS body
  • Power (VAC)100 to 240
  • DescriptionAuto-Desiccator Cabinet
  • Warranty1 year
Reviews
Q&A