Sartorius Arium® RO Cleaning Set
Representative image only.
Sartorius Arium® RO Cleaning Set
Sartorius | Mfr: H2O-CCS| Item: 9929435
Keep your Arium water purification system running smoothly
Call For Price
Call For Price
Buy It Together
Current Item
Sartorius Arium® RO Cleaning Set
Sartorius Arium® RO Cleaning Set
9929435
Call For Price
+
Sartorius Arium Mini Water Purification System
9929310
Call For Price
+
Sartorius Arium® Pro Basic Wall-Mount System
9929340
$9,282.39CAD / Each
+ +
Sartorius Arium® Mini Plus Starter Set
9929444
$12,110.04CAD / Each
+ =
Total Price 34143.87 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Use with Arium Comfort and Advance water systems
  • Effectively remove scaling and metal deposits and eliminate organic compounds
  • 1 year warranty
More About this Item
Filters, cartridges, and other accessories for water purification.
This two-stage cleaning kit removes scaling and organic contaminants. The alkaline cleaning solution dissolves organic compounds while the acidic cleaning solution removes scaling and dissolve metallic deposits.
Specifications
  • Compatible WithArium Comfort and Advance
  • DescriptionCleaning Set for RO Membranes