Sartorius Arium® Pro Basic Wall-Mount System

Representative image only.

Sartorius Arium® Pro Basic Wall-Mount System

Sartorius | Mfr: H2OBASIC-B| Item: 9929340

Each modular designed system focuses on the most important functions to produce high-quality ultrapure water

$9,282.39CAD / Each

$9,282.39CAD / Each

Buy It Together

Current Item

Sartorius Arium® Pro Basic Wall-Mount System
Sartorius Arium® Pro Basic Wall-Mount System
9929340

$9,282.39CAD / Each
+ + =
Total Price 14075.92 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Wall mounting bracket, feed water tubing, rinse water tubing, dispensing tubing, straight connector, screw connector, tubing adapters, and power cord with adapter.

Key Features

  • Each system meets and exceeds the ASTM Type I standard
  • The lowest TOC level, along with a real-time monitoring function
  • Exceptionally retention rates for endotoxins, DNase, and RNase
  • Glass display with touch-activated functions and intuitive menu navigation
  • “Favorites” function for direct access to preprogrammed volume dispensing
  • Order an elemental cartridge kit to complete your system
  • Wall mount to save valuable benchtop space
  • 2 year warranty

More About this Item

The Arium Pro system provides the ultimate flexibility at an excellent cost-benefit ratio while providing the highest quality ultrapure water. Dispense up to 2 L/min of ASTM Type I ultrapure water with conductivity of 0.055 µS/cm (18.2 MO x cm), bacterial content of < 0.001 CFU/mL*, and no particles > 0.22 µm*. Select from five Arium Pro series water purification systems, with models featuring an ultrafilter, UV lamp, or TOC monitor to meet your application needs.

All models feature "Favorite" function for quick access to preprogrammed dispensing volume that saves time and minimizes errors. Graphic and acoustic signals alert you when limits are exceeded or data falls below minimum limits. Easily save your data by connecting to a printer using the RS-232 port.

This wall-mount unit saves valuable space on your lab benchtop. The display and dispensing unit are positioned at the bottom to ensure user-friendly, ergonomic operation. Produced water is great for general laboratory applications such as working with buffer, media, and reagents; histology; ELISA; AAS; and GF-AAS, photometry.

*when using an Arium SterilePlus final filter.

Two year warrantyConformite Europeenne
Specifications
  • ASTM Water GradeType I
  • Capacity40 to 100 L/day
  • Water Purification MethodMixed bed resins and activated carbon, ultrafiltration, UV illumination and sterile filtration
  • Max Flow Rate (L/min)2
  • With UV LampNo
  • With UltrafilterNo
  • TOC MonitorNo
  • SourceDistilled water, DI, RO, or EDI
  • Type I Water (Resistivity)18.2 MΩ × cm at 25°C
  • Type I Water (TOC)< 5 ppb
  • Feed Water Requirements (turbidity)< 1 NTU
  • Type I Water (Conductivity)0.055 µS/cm
  • Type I Water (Bacteria)< 0.001 CFU/mL
  • Feed Water Requirements (pressure)From depressurized atmospheric tank inlet to a maximum inlet pressure of 6.9 (100 psi)
  • Max Inlet Pressure (bar)6.9
  • Power (VAC)100 to 240
  • Height (in)19.4
  • Depth (in)17.75
  • Width (in)13.8
  • Height (cm)49.2
  • Depth (cm)45.1
  • Width (cm)35
  • Special FeaturesWall mount
  • DescriptionPro Basic Wall-Mount System
  • Warranty2 years
Reviews
Q&A