Pomona Electronics 6481 Test Connector Adapter Set, 18 Pieces
Representative image only.
Pomona Electronics 6481 Test Connector Adapter Set, 18 Pieces
Pomona Electronics | Mfr: 6481| Item: 2000945
Available for sale only in the United States and Canada
$192.72CAD / Each
$192.72CAD / Each
Buy It Together
Current Item
Pomona Electronics 6481 Test Connector Adapter Set, 18 Pieces
Pomona Electronics 6481 Test Connector Adapter Set, 18 Pieces
2000945
$192.72CAD / Each
+ =
Total Price 261.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Full set of pin and socket adapters
  • 18 piece set includes one black and red pin and socket of each size
  • Contact sizes: 0.03in, 0.04in, 0.06in, 0.10in
  • Includes Silicone insulated 4ft leads to fit most DMMs from Fluke, Amprobe, Greenlee and others
  • 3 month warranty
3 Month Warranty
Specifications
  • DescriptionTest Connector Adapter Set, 18 Pieces