Pomona Electronics 6723 Test Probes with Screw-on Alligator Clips
Representative image only.
Pomona Electronics 6723 Test Probes with Screw-on Alligator Clips
Pomona Electronics | Mfr: 6723| Item: 2000957
Save time and money with versatile kits
$98.63CAD / Each
$98.63CAD / Each
Product Description
Key Features
  • Probe has a Stainless Steel sharp tip with 8/32 threaded base
  • Eliminates accidental separation with screw on alligator probe tip
  • Test leads fit all standard DMM with safety style banana jacks
  • 3 month warranty
More About this Item
Enhance the versitility of your mulitmeter with these Pomona Test Leads and Kits.
Includes: A set of red and black test leads, and set of red and black medium alligator test clips
3 Month Warranty
Specifications
  • DescriptionTest Probes with Screw-on Alligator Clips