Ohaus Vortex-Genie 504-0234-00 2 Replacement Microcentrifuge Tube Inserts, 2/pk

Representative image only.

Ohaus Vortex-Genie 504-0234-00 2 Replacement Microcentrifuge Tube Inserts, 2/pk

Ohaus | Mfr: 504-0234-00| Item: 0472451

$88.55CAD / Pkg of 2

$88.55CAD / Pkg of 2

Buy It Together

Current Item

Ohaus Vortex-Genie 504-0234-00 2 Replacement Microcentrifuge Tube Inserts, 2/pk
Ohaus Vortex-Genie 504-0234-00 2 Replacement Microcentrifuge Tube Inserts, 2/pk
0472451

$88.55CAD / Pkg of 2
+ + + +
Ohaus Stainless Steel Beads, 5 mm; 100/PK
5140217

$81.48CAD / Each
=
Total Price 2825.08 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 year warranty

More About this Item

Parts and accessories to expand/change capacity and keep your mixer running smoothly.

As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus

1 Year Warranty
Specifications
  • Description2 Replacement Microcentrifuge Tube Inserts, 2/pk
  • Warranty1 year
Reviews
Q&A