Ohaus Vortex-Genie 503-0280-00 Foam Insert for 29-37mm Tubes, Set of 2

Representative image only.

Ohaus Vortex-Genie 503-0280-00 Foam Insert for 29-37mm Tubes, Set of 2

Ohaus | Mfr: 503-0280-00| Item: 0472487

$41.84CAD / Pkg of 2

$41.84CAD / Pkg of 2

Buy It Together

Current Item

Ohaus Vortex-Genie 503-0280-00 Foam Insert for 29-37mm Tubes, Set of 2
Ohaus Vortex-Genie 503-0280-00 Foam Insert for 29-37mm Tubes, Set of 2
0472487

$41.84CAD / Pkg of 2
+ + + +
Ohaus Stainless Steel Beads, 5 mm; 100/PK
5140217

$81.48CAD / Each
=
Total Price 2778.37 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Parts and accessories to expand/change capacity and keep your mixer running smoothly.

As of September 2025 Scientific Industries sold the Vortex Genie product line to Ohaus

Specifications
  • DescriptionFoam Insert for 29-37mm Tubes, Set of 2
Reviews
Q&A