Mettler Toledo U-Electrode Large for Universal Antistatic Kit
Representative image only.
Mettler Toledo U-Electrode Large for Universal Antistatic Kit
Mettler Toledo | Mfr: 63052307| Item: 1133799
Call For Price
Call For Price
Buy It Together
Current Item
Mettler Toledo U-Electrode Large for Universal Antistatic Kit
Mettler Toledo U-Electrode Large for Universal Antistatic Kit
1133799
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Includes
Electrode.
Key Features
  • Can be freely placed next to your balance
  • 3 month warranty
More About this Item
Static and electrostatic charging can cause inaccuracies, greatly affecting sensitive balances like analytical, micro, and semi-micro versions.
Many tare vessels, especially plastic containers, are prone to electrostatic charging. Powdery samples then scatter or stick to the container from the static electricity. If the sample is toxic or harmful, this could be dangerous to the operator.
Discharge/neutralize your samples before weighing to ensure maximum security of the sample and user, and obtain more accurate measurements.
Requires univeral power supply. Does not include a power cord.
3 Month Warranty
Specifications
  • Height (InchShort)8
  • Width (in)7
  • DescriptionU-Electrode Large for Universal Antistatic Kit
  • Warranty3 Months