Mettler Toledo 63052302 Haug Deionizer, with Warning Light
Representative image only.
Mettler Toledo 63052302 Haug Deionizer, with Warning Light
Mettler Toledo | Mfr: 63052302| Item: 1133695
No more scattering of powdery samples
Call For Price
Call For Price
Product Description
Key Features
  • Electrode removes electrostatic discharge reducing contamination and increasing productivity
  • Place electrode next to balance for best results
  • 3 month warranty
More About this Item
This deionizer dissipates static charge from the surrounding area, including your sample. Get accurate, reliable results at a quicker pace. Available with or without a warning light.
3 Month Warranty
Specifications
  • DescriptionHaug Deionizer, with Warning Light