Mettler Toledo NTC 30 kΩ Temperature Compensation Probe, NTC 30 kΩ; 1.2 m Cable, RCA

Representative image only.

Mettler Toledo NTC 30 kΩ Temperature Compensation Probe, NTC 30 kΩ; 1.2 m Cable, RCA

Mettler Toledo | Mfr: 51300164| Item: 3590267

Optimize water analysis

$421.39CAD / Each

$421.39CAD / Each

Buy It Together

Current Item

Mettler Toledo NTC 30 kΩ Temperature Compensation Probe, NTC 30 kΩ; 1.2 m Cable, RCA
Mettler Toledo NTC 30 kΩ Temperature Compensation Probe, NTC 30 kΩ; 1.2 m Cable, RCA
3590267

$421.39CAD / Each
+ + =
Total Price 3181.05 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories
  • Obtain more accurate pH, ISE, mV, conductivity, salinity, and TDS measurements with temperature compensation

More About this Item

For your Mettler Toledo meters and probes.

Specifications
  • Mounting TypeRCA
  • MaterialStainless steel
  • Length (cm)120
  • Width (cm)0.3
  • Height (cm)12
  • DescriptionTemperature Compensation Probe, NTC 30 kΩ; 1.2 m Cable, RCA
Reviews
Q&A