Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 5 m

Representative image only.

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 5 m

Mettler Toledo | Mfr: 30281898| Item: 3590325

Optimize water analysis

Call For Price

Call For Price

Buy It Together

Current Item

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 5 m
Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 5 m
3590325

Call For Price
+ + =
Total Price 2759.66 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories

More About this Item

For your Mettler Toledo meters and probes.

Specifications
  • Length (cm)50
  • DescriptionElectrode Cable, multi-pin to BNC / RCA; 5 m
Reviews
Q&A