Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 3 m

Representative image only.

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 3 m

Mettler Toledo | Mfr: 30281897| Item: 3590324

Optimize water analysis

$356.56CAD / Each

$356.56CAD / Each

Buy It Together

Current Item

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 3 m
Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 3 m
3590324

$356.56CAD / Each
+ + =
Total Price 3116.22 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories

More About this Item

For your Mettler Toledo meters and probes.

Specifications
  • Length (cm)30
  • DescriptionElectrode Cable, multi-pin to BNC / RCA; 3 m
Reviews
Q&A