Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 1.2 m

Representative image only.

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 1.2 m

Mettler Toledo | Mfr: 30281896| Item: 3590323

Optimize water analysis

$282.15CAD / Each

$282.15CAD / Each

Buy It Together

Current Item

Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 1.2 m
Mettler Toledo Inlab® Electrode Cable, multi-pin to BNC / RCA; 1.2 m
3590323

$282.15CAD / Each
+ + =
Total Price 3041.81 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose from a variety of accessories

More About this Item

For your Mettler Toledo meters and probes.

Specifications
  • Length (cm)12
  • DescriptionElectrode Cable, multi-pin to BNC / RCA; 1.2 m
Reviews
Q&A