Manning MSP250 Fixed Speed, Peristaltic, Fluid Sampling Pump
Representative image only.
Manning MSP250 Fixed Speed, Peristaltic, Fluid Sampling Pump
Global Water | Mfr: MS889951-1| Item: 9924300
Ideal for sampling lakes, ponds, holding pools, or shallow wells
$1,672.30CAD / Each
$1,672.30CAD / Each
Buy It Together
Current Item
Manning MSP250 Fixed Speed, Peristaltic, Fluid Sampling Pump
Manning MSP250 Fixed Speed, Peristaltic, Fluid Sampling Pump
9924300
$1,672.30CAD / Each
+ + +
Global Water WS700 Peristaltic Water Sampler
9924370
$4,482.05CAD / Each
+
Manning PSB4A2A2A1F2 Portable Peristaltic Sampler
9924000
$4,724.72CAD / Each
=
Total Price 15018.55 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Simple control for exact sample size
  • Reversible motors for backflushing of sample hose
  • Rugged, UV-protected PVC case with handle for easy use and transportation
  • Flow rate : up to 500 mL/min at 4-ft head, operating temperature: 0 to 70C
  • Features internal 12 VDC rechargeable gel battery and will pump approximately 40 gallons (150 liters) before a recharge is necessary
  • 1 year warranty
More About this Item
These portable sampling pumps are ideal for sampling lakes, ponds, holding pools, or shallow wells. Samplers are packaged in a lightweight, rugged, UV protected PVC case with handle for easy use and transportation. To protect sample integrity the sample only comes in contact with Norprene and polyethylene tubing which is easily cleaned or replaced between samples.
UL Listed to Canadian safety standards.1 Year Warranty
Specifications
  • Power12 VDC rechargable battery (included)
  • DescriptionFixed Speed, Peristaltic, Fluid Sampling Pump