Global Water WS705 Improved Composite/Discrete Water Sampler
Representative image only.
Global Water WS705 Improved Composite/Discrete Water Sampler
Global Water | Mfr: CU1000| Item: 9924375
All the features needed for a wide variety of water sampling requirements
$3,844.07CAD / Each
$3,844.07CAD / Each
Buy It Together
Current Item
Global Water WS705 Improved Composite/Discrete Water Sampler
Global Water WS705 Improved Composite/Discrete Water Sampler
9924375
$3,844.07CAD / Each
+ + + =
Total Price 6730.29 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
15-ft (4.5-m) sampling hose with intake strainer, 1-gallon sample bottle, rechargeable battery, and 120 VAC charger. Model WS755 adds on a second 15-ft (4.5-m) sampling hose and 1-gallon sample bottle.
Key Features
  • Retractable pull handle and heavy-duty wheels provide easy transport
  • Weathertight enclosure for the battery prevents accidental wetness
  • Quick-disconnect pickup hoses can be stored inside the enclosure lid for easy transport
  • 1 year warranty
More About this Item
The WS705/755 samplers combine all of the features you need for stormwater, industrial discharge, water and wastewater, or rivers and stream sampling. Sample size control allows individual time-weighted samples or full-bottle discrete samples. Sample interval controls the time between individual composite samples or enables the external trigger mode. Start delay timer allows multiple units to be set for concurrent sampling or to delay the start of sampling after an event for a more representative sample. An automatic 15-second back-flush cycle cleans debris from the strainer and empties fluid from the hose to prevent contamination.
WS705 sampler features one pump and bottle for most composite/discrete sampling applications. WS755 has two pumps and two bottles, and offers the unique ability to set the sampling mode for each pump and bottle individually. This allows time-weighted composite sample and a discrete sample simultaneously, or you can take two individual time-weighted samples with different sample sizes for each pump.
Order the optional stormwater kit to trigger the sampler for stormwater monitoring applications; includes a rain sensor, flow sensor, and auto-drain rain gauge all in one package.
For full specifications please refer to the instruction manual.
Conformite Europeenne1 Year Warranty
Specifications
  • Sample VolumeAdjustable up to 2.5 gallon
  • Intake Hose TypeReinforced PE (1/4" ID)
  • Bottle TypePolyethylene
  • Sampling PumpPeristaltic
  • TubingReinforced PVC (1/4" ID)
  • Transport Velocity1 LPM at 4-ft head
  • Max Lift (ft)20
  • Max Lift (m)6.1
  • PowerRechargeable battery (included)
  • DescriptionImproved Composite/Discrete Water Sampler