Lightnin 869439 Air-drive mixer with tube suspension

Representative image only.

Lightnin 869439 Air-drive mixer with tube suspension

Lightnin | Mfr: 869439| Item: 5032200

Use these mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical.

$3,167.79CAD / Each

for an additional
$432.44CAD / Each
for an additional
$307.57CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$3,167.79CAD / Each

Buy It Together

Current Item

Lightnin 869439 Air-drive mixer with tube suspension
Lightnin 869439 Air-drive mixer with tube suspension
5032200

$3,167.79CAD / Each
+ +
InnoCal NIST-Traceable Calibration; Mixer
1705003

$307.57CAD / Each
=
Total Price 4593.66 ADD 3 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes. We offer LABMASTER air-drive mixers with two types of mounting styles. The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately). The universal clamp mixers mount directly to a ring stand or tank rim.
Mixers include:
— 5/16" dia x 12"L, 316 SS nonthrough-type shaft
— 5/16" bore x 2.5" dia, 316 SS A-310 impeller
— collet-type chuck
— universal clamp or tube suspension mounting
— muffler
— needle value

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • Material316 Stainless Steel
  • Min Speed (rpm)30
  • Max Speed (rpm)1800
  • Max Torque (in-oz)37
  • Motor TypeAir-driven
  • Max Air Inlet Pressure (PSI)90
  • Min Air Inlet Pressure (PSI)30
  • Shaft Length (in)18
  • Shaft Length (mm)457.2
  • Motor hp1/15
  • DescriptionAir-drive mixer with tube suspension
  • Warranty1 Year
Reviews
Q&A