Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 115V, 30A

Representative image only.

Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 115V, 30A

Labconco | Mfr: 3300600| Item: 3351780

Spacious Class II enclosures provide ample room for a variety of instruments

$68,534.74CAD / Each

$68,534.74CAD / Each

Buy It Together

Current Item

Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 115V, 30A
Labconco Logic® Vue Class II Enclosure, 6-ft Wide; 115V, 30A
3351780

$68,534.74CAD / Each
+ =
Total Price 70634.32 ADD 2 ITEMS TO CART
Product Description

Includes

Four levelling legs with seismic restraint feet, four sash restraint latches, two work surface lift tools, two cable entry 4 to 5 mm inserts, and additional hardware.

Key Features

  • Meets NSF/ANSI 49 requirements for personnel and product protection
  • Continuous airflow monitoring for user safety
  • Two fully opening sashes provide easy equipment loading
  • Ergonomically designed with built-in knee space
  • Spacious working areas: 42.2"H x 31"D (107.2 x 78.7 cm)
  • Durable 304 stainless steel (SS) interior construction
  • Password protection prevents accidental setting changes
  • 6-ft wide enclosure
  • 115 VAC, 50/60 Hz, 30 A
  • 1 year warranty

More About this Item

The Logic® Vue Class II biosafety enclosures are built with your safety and comfort in mind. A HEPA supply and exhaust filters provide >99.99% efficiency (at 0.3 µm) for ISO Class 5/Grade A air or better. Constant Airflow Profile (CAP) system continuously monitors and adjusts airflow to contain biohazards within the enclosure ensuring user safety.

View status information and configure settings using the touchpad controller with a full-color LCD screen. The controller uses MyLogic™ operating system that communicates important status information such as filter life, blower status, alerts, alarms, safety interlock*, and data connectivity. Remotely monitor the status using the USB connection or dry contact relays.

The spacious enclosures accommodate a variety of automated instruments. Multiple pass-throughs allow safe passages of cables, cords, and service fixtures. Durable polycarbonate side windows and fully opening front and rear sashes offer easy accessibility and visibility from all sides. Get unrestricted access to full working area via an 8" (20.3 cm) front lower sash opening. Work surface height is adjustable from 31.5 to 37.5" (80 and 95.3 cm) in 1" increments (2.5 cm); a convenient knee well offers comfort while working seated. A 133 footcandle (1432 lux) LED lighting illuminates work surface for maximum visibility.

Chemically resistant epoxy powder-coated steel exterior and 304 SS interior construction offer excellent durability. A SpillStopper work surface tray accommodates instruments up to 250 lb (113.4 kg) per every 2 feet (61 cm). International packaging models require transportation by high cube shipping containers. Customize your biosafety enclosures with a variety of accessories—order separately.

*Optional accessory
International packaging models require transportation by high cube shipping containers.

UL Listed.Canadian Standards AssociationETL ListedNSF international certified1 Year Warranty
Specifications
  • Blower TypeECM, brushless DC motor
  • Blower (hp)3/4
  • Lights4000 K, 3200 lumens frosted glass
  • Exterior Width (in)78.2
  • Exterior Height (in)99.5
  • Exterior Depth (in)39.6
  • Interior Width (in)73.8
  • Interior Height (in)42.2
  • Interior Depth (in)34.7
  • Power (VAC)115
  • Power (Hz)50/60
  • Sash opening (in)8"
  • Exhaust Volume (CFM)1080 to 1230
  • Shipping Weight (lbs)1275
  • Shipping Weight (kg)578
  • Filter TypeHEPA
  • Number of Filters Required3
  • FixturesWork surface drip tray (measures 71.2"W x 31"D)
  • Description6-ft Wide Class II Enclosure; 115V, 30A
  • Warranty1 Year
Reviews
Q&A