Labconco Fast-Freeze™ 7205101 Ampule Pod, 3/8" ID Tubing; FD
Representative image only.
Labconco Fast-Freeze™ 7205101 Ampule Pod, 3/8" ID Tubing; FD
Labconco | Mfr: 7205101| Item: 0335338
Provides easy and efficient handling of freeze dry samples
$1,399.72CAD / Each
$1,399.72CAD / Each
Buy It Together
Current Item
Labconco Fast-Freeze™ 7205101 Ampule Pod, 3/8" ID Tubing; FD
Labconco Fast-Freeze™ 7205101 Ampule Pod, 3/8" ID Tubing; FD
0335338
$1,399.72CAD / Each
+ + + + + =
Total Price 53351.45 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Includes
15 plugs, 2 clamps, and a straight stainless steel adapter.
Key Features
  • Accommodates up to 15 ampules for simultaneous freeze drying
  • 1 year warranty
More About this Item
The ampule pod is designed to accommodate up to 15 ampules for simultaneous freeze drying. The polypropylene base features fifteen 3/8" ID tubing ports, each with silicone tubing for attachment of ampules (not included). A straight stainless steel adapter easily attaches to a single valve port of a drying chamber, manifold, or freeze drying system.
1 Year Warranty
Specifications
  • Length (")3 7/8
  • Width (InchShort)3 7/8
  • Height (InchShort)7 1/2
  • DescriptionAmpule Pod, 3/8" ID Tubing; FD